missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Telethonin Protein

A DNA sequence encoding the Homo sapiens (Human) Telethonin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6767-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16005690

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_003664.1
Telethonin Protein
Research Use Only
TCAP
Recombinant Protein
E. coli
MATSELSCEV SEENCERREA FWAEWKDLTL STRPEEGCSL HEEDTQRHET YHQQGQCQVL VQRSPWLMMR MGILGRGLQE YQLPYQRVLP LPIFTPAKMG ATKEEREDTP IQLQELLALE TALGGQCVDR QEVAEITKQL PPVVPVSKPG ALRRSLSRSM SQEAQRG
Greater than 80% as determined by SDS-PAGE.
8557
200 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers