missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Surfactant-associated protein 1
A DNA sequence encoding the Homo sapiens (Human) Surfactant-associated protein 3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6680-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001094811.1 | |
Surfactant-associated protein 1 | |
Research Use Only | |
SFTA3 | |
Recombinant Protein | |
E. coli | |
MRAGFSDFQL IRDQVLFLQD QAQRLTEWLQ LSGFENPVSE STTLCLRERE KRIPTCVAVC VPSPGTVHTA LLHPTTLSQS RSSSEAKMLI IHTA | |
Greater than 90% as determined by SDS-PAGE. |
253970 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |