missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Suppressor of SWI4 1 homolog Protein
A DNA sequence encoding the Homo sapiens (Human) Suppressor of SWI4 1 homolog, was expressed in the hosts and tags indicated.
202.00 EUR - 2155.00 EUR
Specifications
Accession Number | NP_001035754.1 |
---|---|
Gene ID (Entrez) | 56342 |
Name | Suppressor of SWI4 1 homolog Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16088941
|
enQuireBio™
QP9895-YE-100UG |
100 μg |
565.00 EUR
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16028951
|
enQuireBio™
QP9895-YE-500UG |
500 μg |
1525.00 EUR
500µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16098941
|
enQuireBio™
QP9895-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16018951
|
enQuireBio™
QP9895-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16038951
|
enQuireBio™
QP9895-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16008951
|
enQuireBio™
QP9895-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001035754.1 | |
Suppressor of SWI4 1 homolog Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
56342 | |
Research Use Only | |
PPAN | |
Recombinant Protein | |
Yeast | |
MGQSGRSRHQ KRARAQAQLR NLEAYAANPH SFVFTRGCTG RNIRQLSLDV RRVMEPLTAS RLQVRKKNSL KDCVAVAGPL GVTHFLILSK TETNVYFKLM RLPGGPTLTF QVKKYSLVRD VVSSLRRHRM HEQQFAHPPL LVLNSFGPHG MHVKLMATMF QNLFPSINVH KVNLNTIKRC LLIDYNPDSQ ELDFRHYSIK VVPVGASRGM KKLLQEKFPN MSRLQDISEL LATGAGLSES EAEPDGDHNI TELPMS-QAV AGRGNMRAQQ SAVRLTEIGP RMTLQLIKVQ EGVGEGKVMF HSFVSKTEEE LQAILEAKEK KLRLKAQRQA QQAQNVQRKQ EQREAHRKKS LEGMKKARVG GSDEEASGIP SRTASLELGE DDDEQEDDDI EYFCQAVGEA PSEDLFPEAK QKRLAKSPGR KRKRWEMDRG RGRLCDQKFP KTKDKSQGAQ ARRGPRGASR DGGRGRGRGR PGKRVA | |
Greater than 90% as determined by SDS-PAGE. |