missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human STC-1 / Stanniocalcin-1 Protein
A DNA sequence encoding the Homo sapiens (Human) STC-1 / Stanniocalcin-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8677-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003146.1 | |
STC-1 / Stanniocalcin-1 Protein | |
Research Use Only | |
STC1 | |
Recombinant Protein | |
E. coli | |
SAEVVRCLNS ALQVGCGAFA CLENSTCDTD GMYDICKSFL YSAAKFDTQG KAFVKESLKC IANGVTSKVF LAIRRCSTFQ RMIAEVQEEC YSKLNVCSIA KRNPEAITEV VQLPNHFSNR YYNRLVRSLL ECDEDTVSTI RDSLMEKIGP NMASLFHILQ TDHCAQTHPR ADFNRRRTNE PQKLKVLLRN LRGEEDSPSH IKRTSHESA | |
Greater than 80% as determined by SDS-PAGE. |
6781 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |