missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human STAT3 Protein
A DNA sequence encoding the Homo sapiens (Human) STAT3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6735-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003141.2 | |
STAT3 Protein | |
Research Use Only | |
STAT3 | |
Recombinant Protein | |
E. coli | |
ESHATLVFHN LLGEIDQQYS RFLQESNVLY QHNLRRIKQF LQSRYLEKPM EIARIVARCL WEESRLLQTA ATAAQQGGQA NHPTAAVVTE KQQMLEQHLQ DVRKRVQDLE QKMKVVENLQ DDFDFNYKTL KSQGDMQDLN GNNQSVTRQK MQQLEQMLTA LDQMRRSIVS ELAGLLSAME YVQKTLTDEE L | |
Greater than 90% as determined by SDS-PAGE. |
6774 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |