missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Src kinase-associated phosphoprotein 1

A DNA sequence encoding the Homo sapiens (Human) Src kinase-associated phosphoprotein 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7785-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16050781

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001068567.1
Src kinase-associated phosphoprotein 1
Research Use Only
SKAP1
Recombinant Protein
E. coli
MQAAALPEEI RWLLEDAEEF LAEGLRNENL SAVARDHRDH ILRGFQQIKA RYYWDFQPQG GDIGQDSSDD NHSGTLGLSL TSDAPFLSDY QDEGMEDIVK GAQELDNVIK QGYLEKKSKD HSFFGSEWQK RWCVVSRGLF YYYANEKSKQ PKGTFLIKGY GVRMAPHLRR DSKKESCFEL TSQDRRSYEF TATSPAEARD WVDQISFLLK DLSSLTIPYE EDEEEEEKEE TYDDIDGFDS PSCGSQCRPT ILPGSVGIKE PTEEKEEEDI YEVLPDEEHD LEEDESGTRR KGVDYASYYQ GLWDCHGDQP DELSFQRGDL IRILSKEYNM YGWWVGELNS LVGIVPKEYL TTAFEVEE
Greater than 90% as determined by SDS-PAGE.
8631
200 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers