missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Spindlin-2B Protein

A DNA sequence encoding the Homo sapiens (Human) Spindlin-2B, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8026-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16042231

  • 680.00 EUR / 100µg
Estimated Shipment: 02-02-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001006682.1
Spindlin-2B Protein
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
474343
Greater than 80% as determined by SDS-PAGE.
Research Use Only
SPIN2B
Recombinant Protein
E. coli
MKTPNAQEAE GQQTRAAAGR ATGSANMTKK KVSQKKQRGR PSSQPRRNIV GCRISHGWKE GDEPITQWKG TVLDQVPINP SLYLVKYDGI DCVYGLELHR DERVLSLKIL SDRVASSHIS DANLANTIIG KAVEHMFEGE HGSKDEWRGM VLAQAPIMKA WFYITYEKDP VLYMYQLLDD YKEGDLRIMP ESSESPPTER EPGGVVDGLI GKHVEYTKED GSKRIGMVIH QVEAKPSVYF IKFDDDFHIY VYDLVKKS
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers