missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Spindlin-2B Protein
A DNA sequence encoding the Homo sapiens (Human) Spindlin-2B, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8026-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001006682.1 | |
Spindlin-2B Protein | |
Research Use Only | |
SPIN2B | |
Recombinant Protein | |
E. coli | |
MKTPNAQEAE GQQTRAAAGR ATGSANMTKK KVSQKKQRGR PSSQPRRNIV GCRISHGWKE GDEPITQWKG TVLDQVPINP SLYLVKYDGI DCVYGLELHR DERVLSLKIL SDRVASSHIS DANLANTIIG KAVEHMFEGE HGSKDEWRGM VLAQAPIMKA WFYITYEKDP VLYMYQLLDD YKEGDLRIMP ESSESPPTER EPGGVVDGLI GKHVEYTKED GSKRIGMVIH QVEAKPSVYF IKFDDDFHIY VYDLVKKS | |
Greater than 80% as determined by SDS-PAGE. |
474343 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |