missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Spike glycoProtein

A DNA sequence encoding the Homo sapiens (Human) Spike glycoprotein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7178-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16027580

  • 458.00 EUR / 100µg
Estimated Shipment: 02-02-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Spike glycoProtein
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Greater than 90% as determined by SDS-PAGE.
Research Use Only
S
Recombinant Protein
E. coli
VIGDLKCTSD NINDKDTGPP PISTDTVDVT NGLGTYYVLD RVYLNTTLFL NGYYPTSGST YRNMALKGSV LLSRLWFKPP FLSDFINGIF AKVKNTKVIK DRVMYSEFPA ITIGSTFVNT SYSVVVQPRT INSTQDGDNK LQGLLEVSVC QYNMCEYPQT ICHPNLGNHR KELWHLDTGV VSCLYKRNFT YDVNADYLYF HFYQEGGTFY AYFTDTGVVT KFLFNVYLGM ALSHYYVMPL TCNSKLTLEY WVTPLTSRQY LLAFNQDGII FNAEDCMSDF MSEIKCKTQS IAPPTGVYEL NGYTVQPIAD VYRRKPNLPN CNIEAWLNDK
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers