missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Spike glycoProtein
A DNA sequence encoding the Homo sapiens (Human) Spike glycoprotein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7178-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
Spike glycoProtein | |
Research Use Only | |
S | |
Recombinant Protein | |
Yeast | |
VIGDLKCTSD NINDKDTGPP PISTDTVDVT NGLGTYYVLD RVYLNTTLFL NGYYPTSGST YRNMALKGSV LLSRLWFKPP FLSDFINGIF AKVKNTKVIK DRVMYSEFPA ITIGSTFVNT SYSVVVQPRT INSTQDGDNK LQGLLEVSVC QYNMCEYPQT ICHPNLGNHR KELWHLDTGV VSCLYKRNFT YDVNADYLYF HFYQEGGTFY AYFTDTGVVT KFLFNVYLGM ALSHYYVMPL TCNSKLTLEY WVTPLTSRQY LLAFNQDGII FNAEDCMSDF MSEIKCKTQS IAPPTGVYEL NGYTVQPIAD VYRRKPNLPN CNIEAWLNDK | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |