missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Smad3 Protein

A DNA sequence encoding the Homo sapiens (Human) Smad3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6697-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16035200

  • 2095.00 EUR / 1mg
Estimated Shipment: 27-01-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001138574.1
Smad3 Protein
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
4088
Greater than 90% as determined by SDS-PAGE.
Research Use Only
SMAD3
Recombinant Protein
E. coli
MSSILPFTPP IVKRLLGWKK GEQNGQEEKW CEKAVKSLVK KLKKTGQLDE LEKAITTQNV NTKCITIPRS LDGRLQVSHR KGLPHVIYCR LWRWPDLHSH HELRAMELCE FAFNMKKDEV CVNPYHYQRV ETPVLPPVLV PRHTEIPAEF PPLDDYSHSI PENTNFPAGI EPQSNIPETP PPGYLSEDGE TSDHQMNHSM DAGSPNLSPN PMSPAHNNLD LQPVTYCEPA FWCSISYYEL NQRVGETFHA SQPSMTVDGF TDPSNSERFC LGLLSNVNRN AAVELTRRHI GRGVRLYYIG GEVFAECLSD SAIFVQSPNC NQRYGWHPAT VCKIPPGCNL KIFNNQEFAA LLAQSVNQGF EAVYQLTRMC TIRMSFVKGW GAEYRRQTVT STPCWIELHL NGPLQWLDKV LTQMGSPSIR CSSVS
Videos
SDS
Documents
Certificates
Special Offers

Special Offers