missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Sjoegren syndrome nuclear autoantigen 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Sjoegren syndrome nuclear autoantigen 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6732-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16035450

  • 463.00 EUR / 100µg
Estimated Shipment: 03-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_003722.2
Sjoegren syndrome nuclear autoantigen 1 Protein
Research Use Only
SSNA1
Recombinant Protein
E. coli
TQQGAALQNY NNELVKCIEE LCQKREELCR QIQEEEDEKQ RLQNEVRQLT EKLARVNENL ARKIASRNEF DRTIAETEAA YLKILESSQT LLSVLKREAG NLTKATAPDQ KSSGGRDS
Greater than 80% as determined by SDS-PAGE.
8636
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers