missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Sjoegren syndrome nuclear autoantigen 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Sjoegren syndrome nuclear autoantigen 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6732-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16075450

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_003722.2
Sjoegren syndrome nuclear autoantigen 1 Protein
Research Use Only
SSNA1
Recombinant Protein
E. coli
TQQGAALQNY NNELVKCIEE LCQKREELCR QIQEEEDEKQ RLQNEVRQLT EKLARVNENL ARKIASRNEF DRTIAETEAA YLKILESSQT LLSVLKREAG NLTKATAPDQ KSSGGRDS
Greater than 80% as determined by SDS-PAGE.
8636
500 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers