missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SerpinA6 / CBG Protein
A DNA sequence encoding the Homo sapiens (Human) SerpinA6 / CBG, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6667-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001747.2 | |
SerpinA6 / CBG Protein | |
Research Use Only | |
SERPINA6 | |
Recombinant Protein | |
Yeast | |
MDPNAAYVNM SNHHRGLASA NVDFAFSLYK HLVALSPKKN IFISPVSISM ALAMLSLGTC GHTRAQLLQG LGFNLTERSE TEIHQGFQHL HQLFAKSDTS LEMTMGNALF LDGSLELLES FSADIKHYYE SEVLAMNFQD WATASRQINS YVKNKTQGKI VDLFSGLDSP AILVLVNYIF FKGTWTQPFD LASTREENFY VDETTVVKVP MMLQSSTISY LHDSELPCQL VQMNYVGNGT VFFILPDKGK MNTVIAALSR DTINRWSAGL TSSQVDLYIP KVTISGVYDL GDVLEEMGIA DLFTNQANFS RITQDAQLKS SKVVHKAVLQ LNEEGVDTAG STGVTLNLTS KPIILRFNQP FIIMIFDHFT WSSLFLARVM NPV | |
Greater than 90% as determined by SDS-PAGE. |
866 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |