missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Selenoprotein 1
A DNA sequence encoding the Homo sapiens (Human) Selenoprotein P, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6665-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001078955.1 | |
Selenoprotein 1 | |
Research Use Only | |
SEPP1 | |
Recombinant Protein | |
E. coli | |
ESQDQSSLCK QPPAWSIRDQ DPMLNSNGSV TVVALLQASS YLCILQASKL EDLRVKLKKE GYSNISYIVV NHQGISSRLK YTHLKNKVSE HIPVYQQEEN QTDVWTLLNG SKDDFLIYDR CGRLVYHLGL PFSFLTFPYV EEAIKIAYCE KKCGNCSLTT LKDEDFCKRV SLATVDKTVE TPSPHYHHEH HHNHGHQHLG SSELSENQQP GAPNAPTHPA PPGLHHHHKH KGQHRQGHPE NRDMPASEDL QDLQKKLCRK RCINQLLCKL PTDSELAPRS SCCHCRHLIF EKTGSAITSQ CKENLPSLCS SQGLRAEENI TESCQSRLPP AASQISQQLI PTEASASSRS KNQAKKSESP SN | |
Greater than 90% as determined by SDS-PAGE. |
6414 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |