missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Secretory phospholipase A2 receptor Protein
A DNA sequence encoding the Homo sapiens (Human) Secretory phospholipase A2 receptor, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7606-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001007268.1 | |
Secretory phospholipase A2 receptor Protein | |
Research Use Only | |
PLA2R1 | |
Recombinant Protein | |
Yeast | |
EEKTWHEALR SCQADNSALI DITSLAEVEF LVTLLGDENA SETWIGLSSN KIPVSFEWSN DSSVIFTNWH TLEPHIFPNR SQLCVSAEQS EGHWKVKNCE ERLFYICKKA GHVLSDAESG CQEGWERHGG FCYKID | |
Greater than 90% as determined by SDS-PAGE. |
22925 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |