missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Scrapie-responsive protein 1

A DNA sequence encoding the Homo sapiens (Human) Scrapie-responsive protein 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7501-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16029030

  • 1525.00 EUR / 500µg

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001316526.1
Scrapie-responsive protein 1
Research Use Only
SCRG1
Recombinant Protein
Yeast
MPANRLSCYR KILKDHNCHN LPEGVADLTQ IDVNVQDHFW DGKGCEMICY CNFSELLCCP KDVFFGPKIS FVIPCNNQ
Greater than 90% as determined by SDS-PAGE.
11341
500 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers