missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SART3 Protein
A DNA sequence encoding the Homo sapiens (Human) SART3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7616-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_055521.1 | |
SART3 Protein | |
Research Use Only | |
SART3 | |
Recombinant Protein | |
E. coli | |
QRKRARAEKK ALKKKKKIRG PEKRGADEDD EKEWGDDEEE QPSKRRRVEN SIPAAGETQN VEVAAGPAGK CAAVDVEPPS KQKEKAASLK RDMPKVLHDS SKDSITVFVS NLPYSMQEPD TKLRPLFEAC GEVVQIRPIF SNRGDFRGYC YVEFKEEKSA LQALEMDRKS VEGRPMFVSP CVDKSKNPDF KVFRYSTSLE KHKLFISGLP FSCTKEELEE ICKAHGTVKD LRLVTNRAGK PKGLAYVEYE NESQASQAVM KMDGMTIKEN IIKVAISNPP QRKVPEKPET RKAPGGPMLL P | |
Greater than 90% as determined by SDS-PAGE. |
9733 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |