missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human S100A15 / S100A7A Protein
A DNA sequence encoding the Homo sapiens (Human) S100A15 / S100A7A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7789-ye-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_789793.1 | |
S100A15 / S100A7A Protein | |
Research Use Only | |
S100A7A | |
Recombinant Protein | |
Yeast | |
SNTQAERSII GMIDMFHKYT GRDGKIEKPS LLTMMKENFP NFLSACDKKG IHYLATVFEK KDKNEDKKID FSEFLSLLGD IAADYHKQSH GAAPCSGGSQ | |
Greater than 90% as determined by SDS-PAGE. |
338324 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |