missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human S100A11 / S100C Protein

A DNA sequence encoding the Homo sapiens (Human) S100A11 / S100C, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6636-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16024710

  • 463.00 EUR / 100µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_005611.1
S100A11 / S100C Protein
Research Use Only
S100A11
Recombinant Protein
E. coli
AKISSPTETE RCIESLIAVF QKYAGKDGYN YTLSKTEFLS FMNTELAAFT KNQKDPGVLD RMMKKLDTNS DGQLDFSEFL NLIGGLAMAC HDSFLKAVPS QKRT
Greater than 90% as determined by SDS-PAGE.
6282
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers