missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human S100A11 / S100C Protein

A DNA sequence encoding the Homo sapiens (Human) S100A11 / S100C, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6636-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16064710

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_005611.1
S100A11 / S100C Protein
Research Use Only
S100A11
Recombinant Protein
E. coli
AKISSPTETE RCIESLIAVF QKYAGKDGYN YTLSKTEFLS FMNTELAAFT KNQKDPGVLD RMMKKLDTNS DGQLDFSEFL NLIGGLAMAC HDSFLKAVPS QKRT
Greater than 90% as determined by SDS-PAGE.
6282
500 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers