missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ryanodine receptor 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Ryanodine receptor 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6633-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000531.2 | |
Ryanodine receptor 1 Protein | |
Research Use Only | |
RYR1 | |
Recombinant Protein | |
E. coli | |
MGDAEGEDEV QFLRTDDEVV LQCSATVLKE QLKLCLAAEG FGNRLCFLEP TSNAQNVPPD LAICCFVLEQ SLSVRALQEM LANTVEAGVE SSQGGGHRTL LYGHAILLRH AHSRMYLSCL TTSRSMTDKL AFDVGLQEDA TGEACWWTMH PASKQRSEGE KVRVGDDIIL VSVSSERYLH LSTASGELQV DASFMQTLWN MNPICSRCEE GFVTGGHVLR LFHGHMDECL TISPADSDDQ RRLVYYEGGA VCTHARSLWR LEPLRISWSG SHLRWGQPLR VRHVTTGQYL ALTEDQGLVV VDASKAHTKA TSFCFRISKE KLDVAPKRDV EGMGPPEIKY GESLCFVQHV ASGLWLTYAA PDPKALRLGV LKKKAMLHQE GHMDDALSLT RCQQEESQAA RMIHSTNGLY NQFIKSLDSF SGKPRGSGPP AGTALPIEGV ILSLQDLIIY FEPPSEDLQH EEKQSKLRSL RNRQSLFQEE GMLSMVLNCI DRLNVYTTAA HFAEFAGEEA AESWKEIVNL LYELLASLIR GNRS | |
Greater than 90% as determined by SDS-PAGE. |
6261 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |