missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RRM1 Protein
A DNA sequence encoding the Homo sapiens (Human) RRM1, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_001024.1 |
---|---|
Gene ID (Entrez) | 6240 |
Name | RRM1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16044660
|
enQuireBio™
QP6629-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16064660
|
enQuireBio™
QP6629-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16054660
|
enQuireBio™
QP6629-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16074660
|
enQuireBio™
QP6629-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16084660
|
enQuireBio™
QP6629-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16094660
|
enQuireBio™
QP6629-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001024.1 | |
RRM1 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
6240 | |
Research Use Only | |
RRM1 | |
Recombinant Protein | |
E. coli | |
MHVIKRDGRQ ERVMFDKITS RIQKLCYGLN MDFVDPAQIT MKVIQGLYSG VTTVELDTLA AETAATLTTK HPDYAILAAR IAVSNLHKET KKVFSDVMED LYNYINPHNG KHSPMVAKST LDIVLANKDR LNSAIIYDRD FSYNYFGFKT LERSYLLKIN GKVAERPQHM LMRVSVGIHK EDIDAAIETY NLLSERWFTH ASPTLFNAGT NRPQLSSCFL LSMKDDSIEG IYDTLKQCAL ISKSAGGIGV AVSCIRATGS YIAGTNGNSN GLVPMLRVYN NTARYVDQGG NKRPGAFAIY LEPWHLDIFE FLDLKKNTGK EEQRARDLFF ALWIPDLFMK RVETNQDWSL MCPNECPGLD EVWGEEFEKL YASYEKQGRV RKVVKAQQLW YAIIESQTET GTPYMLYKDS CNRKSNQQNL GTIKCSNLCT EIVEYTSKDE VAVCNLASLA LNMYVTSEHT YDFKKLAEVT KVVVRNLNKI IDINYYPVPE ACLSNKRHRP IGIGVQGLAD AFILMRYPFE SAEAQLLNKQ IFETIYYGAL EASCDLAKEQ GPYETYEGSP VSKGILQYDM WNVTPTDLWD WKVLKEKIAK YGIRNSLLIA PMPTASTAQI LGNNESIEPY TSNIYTRRVL SGEFQIVNPH LLKDLTERGL WHEEMKNQII ACNGSIQSIP EIPDDLKQLY KTVWEISQKT VLKMAAERGA FIDQSQSLNI HIAEPNYGKL TSMHFYGWKQ GLKTGMYYLR TRPAANPIQF TLNKEKLKDK EKVSKEEEEK ERNTAAMVCS LENRDECLMC GS | |
Greater than 90% as determined by SDS-PAGE. |