missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ROR1 Protein
A DNA sequence encoding the Homo sapiens (Human) ROR1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6615-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001077061.1 | |
ROR1 Protein | |
Research Use Only | |
ROR1 | |
Recombinant Protein | |
Yeast | |
QETELSVSAE LVPTSSWNIS SELNKDSYLT LDEPMNNITT SLGQTAELHC KVSGNPPPTI RWFKNDAPVV QEPRRLSFRS TIYGSRLRIR NLDTTDTGYF QCVATNGKEV VSSTGVLFVK FGPPPTASPG YSDEYEEDGF CQPYRGIACA RFIGNRTVYM ESLHMQGEIE NQITAAFTMI GTSSHLSDKC SQFAIPSLCH YAFPYCDETS SVPKPRDLCR DECEILENVL CQTEYIFARS NPMILMRLKL PNCEDLPQPE SPEAANCIRI GIPMADPINK NHKCYNSTGV DYRGTVSVTK SGRQCQPWNS QYPHTHTFTA LRFPELNGGH SYCRNPGNQK EAPWCFTLDE NFKSDLCDIP AC | |
Greater than 90% as determined by SDS-PAGE. |
4919 | |
200 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |