missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ROR1 Protein

A DNA sequence encoding the Homo sapiens (Human) ROR1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6615-ye-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16054570

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001077061.1
ROR1 Protein
Research Use Only
ROR1
Recombinant Protein
Yeast
QETELSVSAE LVPTSSWNIS SELNKDSYLT LDEPMNNITT SLGQTAELHC KVSGNPPPTI RWFKNDAPVV QEPRRLSFRS TIYGSRLRIR NLDTTDTGYF QCVATNGKEV VSSTGVLFVK FGPPPTASPG YSDEYEEDGF CQPYRGIACA RFIGNRTVYM ESLHMQGEIE NQITAAFTMI GTSSHLSDKC SQFAIPSLCH YAFPYCDETS SVPKPRDLCR DECEILENVL CQTEYIFARS NPMILMRLKL PNCEDLPQPE SPEAANCIRI GIPMADPINK NHKCYNSTGV DYRGTVSVTK SGRQCQPWNS QYPHTHTFTA LRFPELNGGH SYCRNPGNQK EAPWCFTLDE NFKSDLCDIP AC
Greater than 90% as determined by SDS-PAGE.
4919
10 μg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers