missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RING-box protein 1
A DNA sequence encoding the Homo sapiens (Human) RING-box protein 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6613-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001188299.1 | |
RING-box protein 1 | |
Research Use Only | |
RNF7 | |
Recombinant Protein | |
E. coli | |
ADVEDGEETC ALASHSGSSG SKSGGDKMFS LKKWNAVAMW SWDVECDTCA ICRVQVMDAC LRCQAENKQE DCVVVWGECN HSFHNCCMSL WVKQNNRCPL CQQDWVVQRI GK | |
Greater than 90% as determined by SDS-PAGE. |
9616 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |