missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ribosomal protein S6 kinase beta-2 Protein
A DNA sequence encoding the Homo sapiens (Human) Ribosomal protein S6 kinase beta-2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8430-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003943.2 | |
Ribosomal protein S6 kinase beta-2 Protein | |
Research Use Only | |
RPS6KB2 | |
Recombinant Protein | |
E. coli | |
MAAVFDLDLE TEEGSEGEGE PELSPADACP LAELRAAGLE PVGHYEEVEL TETSVNVGPE RIGPHCFELL RVLGKGGYGK VFQVRKVQGT NLGKIYAMKV LRKAKIVRNA KDTAHTRAER NILESVKHPF IVELAYAFQT GGKLYLILEC LSGGELFTHL EREGIFLEDT ACFYLAEITL ALGHLHSQGI IYRDLKPENI MLSSQGHIKL | |
Greater than 90% as determined by SDS-PAGE. |
6199 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |