missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Ribosomal protein S6 kinase beta-2 Protein

A DNA sequence encoding the Homo sapiens (Human) Ribosomal protein S6 kinase beta-2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8430-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16054571

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_003943.2
Ribosomal protein S6 kinase beta-2 Protein
Research Use Only
RPS6KB2
Recombinant Protein
E. coli
MAAVFDLDLE TEEGSEGEGE PELSPADACP LAELRAAGLE PVGHYEEVEL TETSVNVGPE RIGPHCFELL RVLGKGGYGK VFQVRKVQGT NLGKIYAMKV LRKAKIVRNA KDTAHTRAER NILESVKHPF IVELAYAFQT GGKLYLILEC LSGGELFTHL EREGIFLEDT ACFYLAEITL ALGHLHSQGI IYRDLKPENI MLSSQGHIKL
Greater than 90% as determined by SDS-PAGE.
6199
10 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers