missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ribonuclease P protein subunit p29 Protein
A DNA sequence encoding the Homo sapiens (Human) Ribonuclease P protein subunit p29, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6514-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_006618.1 | |
Ribonuclease P protein subunit p29 Protein | |
Research Use Only | |
POP4 | |
Recombinant Protein | |
E. coli | |
MKSVIYHALS QKEANDSDVQ PSGAQRAEAF VRAFLKRSTP RMSPQAREDQ LQRKAVVLEY FTRHKRKEKK KKAKGLSARQ RRELRLFDIK PEQQRYSLFL PLHELWKQYI RDLCSGLKPD TQPQMIQAKL LKADLHGAII SVTKSKCPSY VGITGILLQE TKHIFKIITK EDRLKVIPKL NCVFTVETDG FISYIYGSKF QLRSSERSAK KFKAKGTIDL | |
Greater than 90% as determined by SDS-PAGE. |
10775 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |