missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Retnlb Protein
A DNA sequence encoding the Homo sapiens (Human) Retnlb, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8034-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_115968.1 | |
Retnlb Protein | |
Research Use Only | |
RETNLB | |
Recombinant Protein | |
Yeast | |
QCSLDSVMDK KIKDVLNSLE YSPSPISKKL SCASVKSQGR PSSCPAGMAV TGCACGYGCG SWDVQLETTC HCQCSVVDWT TARCCHLT | |
Greater than 90% as determined by SDS-PAGE. |
84666 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |