missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Retinoschisin Protein
A DNA sequence encoding the Homo sapiens (Human) Retinoschisin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6631-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000321.1 | |
Retinoschisin Protein | |
Research Use Only | |
RS1 | |
Recombinant Protein | |
E. coli | |
STEDEGEDPW YQKACKCDCQ GGPNALWSAG ATSLDCIPEC PYHKPLGFES GEVTPDQITC SNPEQYVGWY SSWTANKARL NSQGFGCAWL SKFQDSSQWL QIDLKEIKVI SGILTQGRCD IDEWMTKYSV QYRTDERLNW IYYKDQTGNN RVFYGNSDRT STVQNLLRPP IISRFIRLIP LGWHVRIAIR MELLECVSKC A | |
Greater than 90% as determined by SDS-PAGE. |
6247 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |