missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Retinal dehydrogenase 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Retinal dehydrogenase 2, was expressed in the hosts and tags indicated.
287.00 EUR - 3175.00 EUR
Specifications
Accession Number | NP_001193826.1 |
---|---|
Gene ID (Entrez) | 8854 |
Name | Retinal dehydrogenase 2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16027331
|
enQuireBio™
QP8968-YE-100UG |
100 μg |
810.00 EUR
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16077331
|
enQuireBio™
QP8968-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16037331
|
enQuireBio™
QP8968-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16057331
|
enQuireBio™
QP8968-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16067331
|
enQuireBio™
QP8968-YE-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16047331
|
enQuireBio™
QP8968-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001193826.1 | |
Retinal dehydrogenase 2 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
8854 | |
Research Use Only | |
ALDH1A2 | |
Recombinant Protein | |
Yeast | |
MTSSKIEMPG EVKADPAALM ASLHLLPSPT PNLEIKYTKI FINNEWQNSE SGRVFPVYNP ATGEQVCEVQ EADKADIDKA VQAARLAFSL GSVWRRMDAS ERGRLLDKLA DLVERDRAVL ATMESLNGGK PFLQAFYVDL QGVIKTFRYY AGWADKIHGM TIPVDGDYFT FTRHEPIGVC GQIIPWNFPL LMFAWKIAPA LCCGNTVVIK PAEQTPLSAL YMGALIKEAG FPPGVINILP GYGPTAGAAI ASHIGIDKIA FTGSTEVGKL IQEAAGRSNL KRVTLELGGK SPNIIFADAD LDYAVEQAHQ GVFFNQGQCC TAGSRIFVEE SIYEEFVRRS VERAKRRVVG SPFDPTTEQG PQIDKKQYNK ILELIQSGVA EGAKLECGGK GLGRKGFFIE PTVFSNVTDD MRIAKEEIFG PVQEILRFKT MDEVIERANN SDFGLVAAVF TNDINKALTV SSAMQAGTVW INCYNALNAQ SPFGGFKMSG NGREMGEFGL REYSEVKTVT VKIPQKNS | |
Greater than 90% as determined by SDS-PAGE. |