missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Replication protein A 14 kDa subunit Protein
A DNA sequence encoding the Homo sapiens (Human) Replication protein A 14 kDa subunit, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8853-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_002938.1 | |
Replication protein A 14 kDa subunit Protein | |
Research Use Only | |
RPA3 | |
Recombinant Protein | |
E. coli | |
MVDMMDLPRS RINAGMLAQF IDKPVCFVGR LEKIHPTGKM FILSDGEGKN GTIELMEPLD EEISGIVEVV GRVTAKATIL CTSYVQFKED SHPFDLGLYN EAVKIIHDFP QFYPLGIVQ | |
Greater than 90% as determined by SDS-PAGE. |
6119 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |