missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Regulator of G-protein signaling 7 Protein
A DNA sequence encoding the Homo sapiens (Human) Regulator of G-protein signaling 7, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6606-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001269702.1 | |
Regulator of G-protein signaling 7 Protein | |
Research Use Only | |
RGS7 | |
Recombinant Protein | |
E. coli | |
MAQGNNYGQT SNGVADESPN MLVYRKMEDV IARMQDEKNG IPIRTVKSFL SKIPSVFSGS DIVQWLIKNL TIEDPVEALH LGTLMAAHGY FFPISDHVLT LKDDGTFYRF QTPYFWPSNC WEPENTDYAV YLCKRTMQNK ARLELADYEA ESLARLQRAF ARKWEFIFMQ AEAQAKVDKK RDKIERKILD SQERAFWDVH RPVPGCVNTT EVDIKKSSRM RNPHKTRKSV YGLQNDIRSH SPTHTPTPET KPPTEDELQQ QIKYWQIQLD RHRLKMSKVA DSLLSYTEQY LEYDPFLLPP DPSNPWLSDD TTFWELEASK EPSQQRVKRW GFGMDEALKD PVGREQFLKF LESEFSSENL RFWLAVEDLK KRPIKEVPSR VQEIWQEFLA PGAPSAINLD SKSYDKTTQN VKEPGRYTFE DAQEHIYKLM KSDSYPRFIR SSAYQELLQA KKKSGNSMDR RTSFEKFAQN VGKSLTSKRL TSLAQSY | |
Greater than 90% as determined by SDS-PAGE. |
6000 | |
100 μg | |
Not determined. | |
Full Length of Isoform 5 | |
Human | |
His | |
Tris-based buffer, 50% glycerol |