missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Recoverin Protein

A DNA sequence encoding the Homo sapiens (Human) Recoverin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6591-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16024380

  • 458.00 EUR / 100µg
Estimated Shipment: 02-02-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_002894.1
Recoverin Protein
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
5957
Greater than 90% as determined by SDS-PAGE.
Research Use Only
RCVRN
Recombinant Protein
E. coli
GNSKSGALSK EILEELQLNT KFSEEELCSW YQSFLKDCPT GRITQQQFQS IYAKFFPDTD PKAYAQHVFR SFDSNLDGTL DFKEYVIALH MTTAGKTNQK LEWAFSLYDV DGNGTISKNE VLEIVMAIFK MITPEDVKLL PDDENTPEKR AEKIWKYFGK NDDDKLTEKE FIEGTLANKE ILRLIQFEPQ KVKEKMKNA
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers