missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Ras-related protein Rab-4A Protein

A DNA sequence encoding the Homo sapiens (Human) Ras-related protein Rab-4A, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6570-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16094240

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001258927.1
Ras-related protein Rab-4A Protein
Research Use Only
RAB4A
Recombinant Protein
E. coli
MSQTAMSETY DFLFKFLVIG NAGTGKSCLL HQFIEKKFKD DSNHTIGVEF GSKIINVGGK YVKLQIWDTA GQERFRSVTR SYYRGAAGAL LVYDITSRET YNALTNWLTD ARMLASQNIV IILCGNKKDL DADREVTFLE ASRFAQENEL MFLETSALTG ENVEEAFVQC ARKILNKIES GELDPERMGS GIQYGDAALR QLRSPRRAQA PNAQECGC
Greater than 80% as determined by SDS-PAGE.
5867
50 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers