missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RAB27B Protein
A DNA sequence encoding the Homo sapiens (Human) RAB27B, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_004154.2 |
---|---|
Gene ID (Entrez) | 5874 |
Name | RAB27B Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16024230
|
enQuireBio™
QP6568-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16044230
|
enQuireBio™
QP6568-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16034230
|
enQuireBio™
QP6568-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16054230
|
enQuireBio™
QP6568-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16064230
|
enQuireBio™
QP6568-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16074230
|
enQuireBio™
QP6568-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_004154.2 | |
RAB27B Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
5874 | |
Research Use Only | |
RAB27B | |
Recombinant Protein | |
E. coli | |
TDGDYDYLIK LLALGDSGVG KTTFLYRYTD NKFNPKFITT VGIDFREKRV VYNAQGPNGS SGKAFKVHLQ LWDTAGQERF RSLTTAFFRD AMGFLLMFDL TSQQSFLNVR NWMSQLQANA YCENPDIVLI GNKADLPDQR EVNERQAREL ADKYGIPYFE TSAATGQNVE KAVETLLDLI MKRMEQCVEK TQIPDTVNGG NSGNLDGEKP PEKKCIC | |
Greater than 90% as determined by SDS-PAGE. |