missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Pyridoxal kinase Protein

A DNA sequence encoding the Homo sapiens (Human) Pyridoxal kinase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6481-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16023620

  • 2095.00 EUR / 1mg
Estimated Shipment: 30-01-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_003672.1
Pyridoxal kinase Protein
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
8566
Greater than 80% as determined by SDS-PAGE.
Research Use Only
PDXK
Recombinant Protein
E. coli
MEEECRVLSI QSHVIRGYVG NRAATFPLQV LGFEIDAVNS VQFSNHTGYA HWKGQVLNSD ELQELYEGLR LNNMNKYDYV LTGYTRDKSF LAMVVDIVQE LKQQNPRLVY VCDPVLGDKW DGEGSMYVPE DLLPVYKEKV VPLADIITPN QFEAELLSGR KIHSQEEALR VMDMLHSMGP DTVVITSSDL PSPQGSNYLI VLGSQRRRNP AGSVVMERIR MDIRKVDAVF VGTGDLFAAM LLAWTHKHPN NLKVACEKTV STLHHVLQRT IQCAKAQAGE GVRPSPMQLE LRMVQSKRDI EDPEIVVQAT VL
Videos
SDS
Documents
Certificates
Special Offers

Special Offers