missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Pyridoxal kinase Protein
A DNA sequence encoding the Homo sapiens (Human) Pyridoxal kinase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6481-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003672.1 | |
Pyridoxal kinase Protein | |
Research Use Only | |
PDXK | |
Recombinant Protein | |
E. coli | |
MEEECRVLSI QSHVIRGYVG NRAATFPLQV LGFEIDAVNS VQFSNHTGYA HWKGQVLNSD ELQELYEGLR LNNMNKYDYV LTGYTRDKSF LAMVVDIVQE LKQQNPRLVY VCDPVLGDKW DGEGSMYVPE DLLPVYKEKV VPLADIITPN QFEAELLSGR KIHSQEEALR VMDMLHSMGP DTVVITSSDL PSPQGSNYLI VLGSQRRRNP AGSVVMERIR MDIRKVDAVF VGTGDLFAAM LLAWTHKHPN NLKVACEKTV STLHHVLQRT IQCAKAQAGE GVRPSPMQLE LRMVQSKRDI EDPEIVVQAT VL | |
Greater than 80% as determined by SDS-PAGE. |
8566 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |