Learn More
enQuireBio™ Recombinant Human Pyridoxal kinase Protein
A DNA sequence encoding the Homo sapiens (Human) Pyridoxal kinase, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP6481-ec-100ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16003620
  • 525.00 EUR / 100µg
Estimated Shipment: 26-07-2024
Specifications
NP_003672.1
Pyridoxal kinase Protein
Research Use Only
PDXK
Recombinant Protein
E. coli
MEEECRVLSI QSHVIRGYVG NRAATFPLQV LGFEIDAVNS VQFSNHTGYA HWKGQVLNSD ELQELYEGLR LNNMNKYDYV LTGYTRDKSF LAMVVDIVQE LKQQNPRLVY VCDPVLGDKW DGEGSMYVPE DLLPVYKEKV VPLADIITPN QFEAELLSGR KIHSQEEALR VMDMLHSMGP DTVVITSSDL PSPQGSNYLI VLGSQRRRNP AGSVVMERIR MDIRKVDAVF VGTGDLFAAM LLAWTHKHPN NLKVACEKTV STLHHVLQRT IQCAKAQAGE GVRPSPMQLE LRMVQSKRDI EDPEIVVQAT VL
Greater than 80% as determined by SDS-PAGE.
8566
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol