missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PTPRN Protein
A DNA sequence encoding the Homo sapiens (Human) PTPRN, was expressed in the hosts and tags indicated.
202.00 EUR - 2155.00 EUR
Specifications
Accession Number | NP_001186692.1 |
---|---|
Gene ID (Entrez) | 5798 |
Name | PTPRN Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16028641
|
enQuireBio™
QP9748-YE-100UG |
100 μg |
565.00 EUR
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16068641
|
enQuireBio™
QP9748-YE-500UG |
500 μg |
1525.00 EUR
500µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16038641
|
enQuireBio™
QP9748-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16058641
|
enQuireBio™
QP9748-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16078641
|
enQuireBio™
QP9748-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16048641
|
enQuireBio™
QP9748-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001186692.1 | |
PTPRN Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
5798 | |
Research Use Only | |
PTPRN | |
Recombinant Protein | |
Yeast | |
VSAHGCLFDR RLCSHLEVCI QDGLFGQCQV GVGQARPLLQ VTSPVLQRLQ GVLRQLMSQG LSWHDDLTQY VISQEMERIP RLRPPEPRPR DRSGLAPKRP GPAGELLLQD IPTGSAPAAQ HRLPQPPVGK GGAGASSSLS PLQAELLPPL LEHLLLPPQP PHPSLSYEPA LLQPYLFHQF GSRDGSRVSE GSPGMVSVGP LPKAEAPALF SRTASKGIFG DHPGHSYGDL PGPSPAQLFQ DSGLLYLAQE LPAPSRARVP RLPEQGSSSR AEDSPEGYEK EGLGDRGEKP ASPAVQPDAA LQRLAAVLAG YGVELRQLTP EQLSTLLTLL QLLPKGAGRN PGGVVNVGAD IKKTMEGPVE GRDTAELPAR TSPMPGHPTA SPTSSEVQQV PSPVSSEPPK AARPPVTPVL LEKKSPLGQS QPTVAGQPSA RPAAEEYGYI VTDQKPLSLA AGVKLLEILA EHVHMSSGSF INISVVGPAL TFRIRHNEQN LSLADVTQQA GLVKSELEAQ TGLQILQTGV GQREEAAAVL PQTAHSTSPM R | |
Greater than 90% as determined by SDS-PAGE. |