missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Pterin-4-alpha-carbinolamine dehydratase Protein
A DNA sequence encoding the Homo sapiens (Human) Pterin-4-alpha-carbinolamine dehydratase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9320-ye-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000272.1 | |
Pterin-4-alpha-carbinolamine dehydratase Protein | |
Research Use Only | |
PCBD1 | |
Recombinant Protein | |
Yeast | |
AGKAHRLSAE ERDQLLPNLR AVGWNELEGR DAIFKQFHFK DFNRAFGFMT RVALQAEKLD HHPEWFNVYN KVHITLSTHE CAGLSERDIN LASFIEQVAV SMT | |
Greater than 90% as determined by SDS-PAGE. |
5092 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |