missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein yippee-like 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Protein yippee-like 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6896-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_037445.1 | |
Protein yippee-like 1 Protein | |
Research Use Only | |
YPEL1 | |
Recombinant Protein | |
E. coli | |
MVKMTKSKTF QAYLPNCHRT YSCIHCRAHL ANHDELISKS FQGSQGRAYL FNSVVNVGCG PAEERVLLTG LHAVADIYCE NCKTTLGWKY EHAFESSQKY KEGKFIIELA HMIKDNGWE | |
Greater than 90% as determined by SDS-PAGE. |
29799 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |