Learn More
enQuireBio™ Recombinant Human Protein yippee-like 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Protein yippee-like 1, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP6896-ec-1mg
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16056620
  • 2090.00 EUR / 1mg
Estimated Shipment: 04-07-2024
Specifications
NP_037445.1
Protein yippee-like 1 Protein
Research Use Only
YPEL1
Recombinant Protein
E. coli
MVKMTKSKTF QAYLPNCHRT YSCIHCRAHL ANHDELISKS FQGSQGRAYL FNSVVNVGCG PAEERVLLTG LHAVADIYCE NCKTTLGWKY EHAFESSQKY KEGKFIIELA HMIKDNGWE
Greater than 90% as determined by SDS-PAGE.
29799
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol