missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein Wnt-4 Protein
A DNA sequence encoding the Homo sapiens (Human) Protein Wnt-4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6888-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_110388.2 | |
Protein Wnt-4 Protein | |
Research Use Only | |
WNT4 | |
Recombinant Protein | |
E. coli | |
SNWLYLAKLS SVGSISEEET CEKLKGLIQR QVQMCKRNLE VMDSVRRGAQ LAIEECQYQF RNRRWNCSTL DSLPVFGKVV TQGTREAAFV YAISSAGVAF AVTRACSSGE LEKCGCDRTV HGVSPQGFQW SGCSDNIAYG VAFSQSFVDV RERSKGASSS RALMNLHNNE AGRKAILTHM RVECKCHGVS GSCEVKTCWR AVPPFRQVGH ALKEKFDGAT EVEPRRVGSS RALVPRNAQF KPHTDEDLVY LEPSPDFCEQ DMRSGVLGTR GRTCNKTSKA IDGCELLCCG RGFHTAQVEL AERCSCKFHW CCFVKCRQCQ RLVELHTCR | |
Greater than 90% as determined by SDS-PAGE. |
54361 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |