missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Protein quaking Protein

A DNA sequence encoding the Homo sapiens (Human) Protein quaking, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7918-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16031581

  • 463.00 EUR / 100µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001288014.1
Protein quaking Protein
Research Use Only
QKI
Recombinant Protein
E. coli
MVGEMETKEK PKPTPDYLMQ LMNDKKLMSS LPNFCGIFNH LERLLDEEIS RVRKDMYNDT LNGSTEKRSA ELPDAVGPIV QLQEKLYVPV KEYPDFNFVG RILGPRGLTA KQLEAETGCK IMVRGKGSMR DKKKEEQNRG KPNWEHLNED LHVLITVEDA QNRAEIKLKR AVEEVKKLLV PAAEGEDSLK KMQLMELAIL NGTYRDANIK SPALAFSLAA TAQAAPRIIT GPAPVLPPAA LRTPTPAGPT IMPLIRQIQT AVMPNGTPHP TAAIVPPGPE AGLIYTPYEY PYTLAPATSI LEYPIEPSGV LGAVATKVRR HDMRVHPYQR IVTADRAATG N
Greater than 80% as determined by SDS-PAGE.
9444
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers