missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Protein E7 Protein

A DNA sequence encoding the Homo sapiens (Human) Protein E7, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7199-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16097660

  • 463.00 EUR / 100µg
Estimated Shipment: 26-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Protein E7 Protein
Research Use Only
E7
Recombinant Protein
E. coli
MRGDKATIKD YILDLQPETT DLHCYEQLGD SSDEEDTDGV DRPDGQAEQA TSNYYIVTYC HSCDSTLRLC IHSTATDLRT LQQMLLGTLQ VVCPGCARL
Greater than 90% as determined by SDS-PAGE.
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers