missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein E7 Protein
A DNA sequence encoding the Homo sapiens (Human) Protein E7, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7199-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
Protein E7 Protein | |
Research Use Only | |
E7 | |
Recombinant Protein | |
E. coli | |
MRGDKATIKD YILDLQPETT DLHCYEQLGD SSDEEDTDGV DRPDGQAEQA TSNYYIVTYC HSCDSTLRLC IHSTATDLRT LQQMLLGTLQ VVCPGCARL | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |