missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein E6 Protein
A DNA sequence encoding the Homo sapiens (Human) Protein E6, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7166-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
Protein E6 Protein | |
Research Use Only | |
E6 | |
Recombinant Protein | |
E. coli | |
MFEDPATRPR TLHELCEVLE ESVHEIRLQC VQCKKELQRR EVYKFLFTDL RIVYRDNNPY GVCIMCLRFL SKISEYRHYQ YSLYGKTLEE RVKKPLSEIT IRCIICQTPL CPEEKERHVN ANKRFHNIMG RWTGRCSECW RPRPVTQV | |
Greater than 90% as determined by SDS-PAGE. |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |