missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein boule-like Protein
A DNA sequence encoding the Homo sapiens (Human) Protein boule-like, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7988-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001271287.1 | |
Protein boule-like Protein | |
Research Use Only | |
BOLL | |
Recombinant Protein | |
E. coli | |
MQTDSLSPSP NPVSPVPLNN PTSAPRYGTV IPNRIFVGGI DFKTNESDLR KFFSQYGSVK EVKIVNDRAG VSKGYGFVTF ETQEDAQKIL QEAEKLNYKD KKLNIGPAIR KQQVGIPRSS IMPAAGTMYL TTSTGYPYTY HNGVAYFHTP EVTSVPPPWP SRSVCSSPVM VAQPIYQQPA YHYQATTQYL PGQWQWSVPQ PSASSAPFLY LQPSEVIYQP VEIAQDGGCV PPPLSLMETS VPEPYSDHGV QATYHQVYAP SAITMPAPVM QPEPIKTVWS IHY | |
Greater than 80% as determined by SDS-PAGE. |
66037 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |